3-Methoxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: Vanillin 4-O-b-D-Glucoside, Glucovanillin

A323418-250MG A323418-1G
103825-340EA 470 USD
103825-340 76802-932 76839-194
3-Methoxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzaldehyde 95%
3-Methoxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₈O₈
MW: 314.29 g/mol
Boiling Pt: 574.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00210591
CAS Number: 494-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR