2-Methyl-m-anisic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxy-2-methylbenzoic acid

A311507-25G A311507-1000G A311507-100G A311507-10G A311507-500G A311507-5G
103835-860EA 5 USD
103835-860 76838-664 77250-984 77250-986 77250-988 77250-990 77306-056
2-Methyl-m-anisic acid 98%
2-Methyl-m-anisic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 297 °C (760 mm Hg)
Flash Pt: 119.9 °C
Storage Temperature: Ambient
MDL Number: MFCD02094039
CAS Number: 55289-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR