3-Methoxy-2,2-dimethyl-3-oxopropanoic acid 91%

Supplier: Ambeed
Warning

A390469-1G A390469-25G A390469-5G A390469-100MG A390469-250MG
103826-862EA 28 USD
103826-862 103837-904 76841-290 76841-292 76841-294 77252-016 77252-018
3-Methoxy-2,2-dimethyl-3-oxopropanoic acid 91%
3-Methoxy-2,2-dimethyl-3-oxopropanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₄
MW: 146.141 g/mol
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD09833517
CAS Number: 13051-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR