3-Mercapto-3-methylbutyl formate 95%

Supplier: Ambeed
Danger

A265815-1G A265815-250MG
103812-760EA 35 USD
103812-760 103852-656 77304-934 77685-759
3-Mercapto-3-methylbutyl formate 95%
3-Mercapto-3-methylbutyl formate

Precaución: for Research Use Only

Formula: C₆H₁₂O₂S
Boiling Pt: 195.5 °C (760 mm Hg)
Flash Pt: 77.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08457872
CAS Number: 50746-10-6
UN: 3334
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR