3-Maleimidopropionic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid, 3-Maleimido-propionic acid, N-(2-Carboxyethyl)maleimide, MPS-Acid, N-Maleoyl-β-alanine
A114517-100G
A114517-10G
A114517-250MG
A114517-1G
A114517-5G
A114517-25G
103798-420EA
35
USD
103798-420
103824-260
77094-184
77094-186
77094-188
77311-074
77311-076
77549-236
3-Maleimidopropionic acid 95%
3-Maleimidopropionic acid
Precaución: for Research Use Only
Formula:
C₇H₇NO₄ MW: 169.14 g/mol Melting Pt: 103…106 °C Flash Pt: 185.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043030 CAS Number: 7423-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...