Isoquinoline-3-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Isoquinolinecarbonitrile

A107424-1G A107424-5G A107424-10G A107424-250MG
103797-186EA 19 USD
103797-186 103822-956 77309-804 77309-806 77570-452 77670-534
Isoquinoline-3-carbonitrile 98%
Isoquinoline-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MW: 154.17 g/mol
Boiling Pt: 348.9 °C (760 mm Hg)
Melting Pt: 126…128 °C
Flash Pt: 121.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00075136
CAS Number: 26947-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR