3-Isopropylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Isopropylbenzeneboronic acid

A102346-10G A102346-250MG A102346-25G A102346-1G A102346-5G
103796-360EA 12 USD
103796-360 103822-052 77245-176 77245-178 77245-180 77282-924 77282-926
3-Isopropylphenylboronic acid 98%
3-Isopropylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
Boiling Pt: 294.2 °C (760 mm Hg)
Melting Pt: 75…80 °C
Flash Pt: 131.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01074634
CAS Number: 216019-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR