3-(Propan-2-yloxy)azetidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Isopropoxyazetidine hydrochloride

A133632-10G A133632-250MG A133632-25G A133632-1G A133632-5G
103801-890EA 20 USD
103801-890 76983-854 76983-856 76983-858 77298-642 77298-644
3-(Propan-2-yloxy)azetidine hydrochloride 97%
3-(Propan-2-yloxy)azetidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD06804558
CAS Number: 871657-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR