3-Isobutylglutaric acid 98%

Supplier: Ambeed
Warning

A186579-100G A186579-10G A186579-500G A186579-1000G A186579-25G
103809-204EA 176 USD
103809-204 76974-662 76974-664 76974-666 77084-196 77084-198
3-Isobutylglutaric acid 98%
3-Isobutylglutaric acid

Precaución: for Research Use Only

Formula: C₉H₁₆O₄
Boiling Pt: 326.2 °C (760 mm Hg)
Flash Pt: 165.289 °C
Storage Temperature: Freezer
MDL Number: MFCD15529751
CAS Number: 75143-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR