3-Iodothiophene 97%

Supplier: Ambeed
Warning

A148377-100G A148377-10G A148377-1G A148377-25G A148377-5G
103803-924EA 5 USD
103803-924 103842-036 77272-866 77272-868 77317-496 77317-498 77317-500
3-Iodothiophene 97%
3-Iodothiophene

Precaución: for Research Use Only

Formula: C₄H₃IS
Boiling Pt: 195.2 °C (760 mm Hg)
Melting Pt: 13 °C
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037901
CAS Number: 10486-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR