3-Iodopyridine 98%
Supplier: Ambeed
|
Warning
|
A176020-1G
A176020-25G
A176020-5G
A176020-100G
A176020-10G
A176020-250MG
A176020-500G
103807-110EA
6
USD
103807-110
103845-422
77082-770
77082-772
77082-774
77287-002
77287-004
77287-006
77287-008
3-Iodopyridine 98%
3-Iodopyridine
Precaución: for Research Use Only
Formula:
C₅H₄IN Melting Pt: 53…56 °C Flash Pt: 106.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023553 CAS Number: 1120-90-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...