3-Iodo-2-propynyl-N-butylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Iodoprop-2-yn-1-yl butylcarbamate, N-Butylcarbamic Acid 3-Iodo-2-propynyl Ester, IPBC

A314951-25G A314951-100G A314951-10G
103835-976EA 6 USD
103835-976 76838-806 77194-740 77194-742
3-Iodo-2-propynyl-N-butylcarbamate 98%
3-Iodo-2-propynyl-N-butylcarbamate

Precaución: for Research Use Only

Formula: C₈H₁₂INO₂
MW: 281.09 g/mol
Boiling Pt: 321.8 °C (760 mm Hg)
Melting Pt: 64…68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00072438
CAS Number: 55406-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR