3-Iodo-9-phenylcarbazole 98%

Supplier: Ambeed
Warning

A142976-100G A142976-1MG A142976-1G A142976-25G A142976-5G
103803-230EA 6 USD
103803-230 77272-562 77272-564 77299-320 77316-474 77316-476
3-Iodo-9-phenylcarbazole 98%
3-Iodo-9-phenylcarbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₂IN
Boiling Pt: 480.6 °C (760 mm Hg)
Flash Pt: 244.5 °C
Storage Temperature: Ambient
MDL Number: MFCD09261276
CAS Number: 502161-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR