3-Iodobenzylamine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A306891-1G
A306891-25G
A306891-10G
A306891-250MG
A306891-100G
103814-484EA
19
USD
103814-484
103835-656
76838-386
76838-388
77277-428
77277-430
77550-840
3-Iodobenzylamine hydrochloride 97%
3-Iodobenzylamine hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₉ClIN MW: 269.5105 g/mol Boiling Pt: 272.5 °C (760 mm Hg) Melting Pt: 188…190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012857 CAS Number: 3718-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...