3-Iodoaniline 98%

Supplier: Ambeed
Warning

A231130-25G A231130-5G A231130-100G A231130-10G
103850-500EA 5 USD
103850-500 77089-376 77089-378 77113-008 77113-010
3-Iodoaniline 98%
3-Iodoaniline

Precaución: for Research Use Only

Formula: C₆H₆IN
MW: 219.02 g/mol
Boiling Pt: 271.6 °C (760 mm Hg)
Melting Pt: 21…24 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007781
CAS Number: 626-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR