3-Iodo-7-azaindole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A519491-250MG A519491-1G
103828-490EA 78 USD
103828-490 77156-862 77307-708
3-Iodo-7-azaindole-5-carboxylic acid 95%
3-Iodo-7-azaindole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅IN₂O₂
MW: 288.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10698484
CAS Number: 1060816-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR