3-Iodo-5-methyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

A116767-100MG A116767-250MG A116767-1G A116767-5G
103798-792EA 36 USD
103798-792 103824-660 76822-138 76822-140 77311-456 77311-458
3-Iodo-5-methyl-1H-pyrazole 97%
3-Iodo-5-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅IN₂
Storage Temperature: Refrigerator
MDL Number: MFCD23135760
CAS Number: 93233-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR