3-Iodo-5-methoxybenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A677568-250MG
A677568-1G
103840-526EA
571
USD
103840-526
77104-772
77308-114
3-Iodo-5-methoxybenzoic acid 95%
3-Iodo-5-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇IO₃ MW: 278.04 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD21607480 CAS Number: 74440-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...