3-Iodo-4-nitropyridine 97%

Supplier: Ambeed
Warning

A121700-250MG A121700-1G
103800-112EA 136 USD
103800-112 77271-538 77312-240
3-Iodo-4-nitropyridine 97%
3-Iodo-4-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃IN₂O₂
MW: 249.99 g/mol
Boiling Pt: 313.479 °C (760 mm Hg)
Flash Pt: 143.388 °C
Storage Temperature: Ambient
MDL Number: MFCD09835271
CAS Number: 89283-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR