3-Iodo-4-nitrophenol 97%

Supplier: Ambeed
Danger

A116206-250MG A116206-1G A116206-5G
103798-696EA 34 USD
103798-696 103824-558 76971-190 77311-376 77311-378
3-Iodo-4-nitrophenol 97%
3-Iodo-4-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄INO₃
Boiling Pt: 382.5 °C (760 mm Hg)
Melting Pt: 153…159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11617270
CAS Number: 50590-07-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR