3-Iodo-p-toluic acid 98%

Supplier: Ambeed
Warning

A142593-100G A142593-10G A142593-25G A142593-5G
77272-536EA 67 USD
77272-536 77272-538 77316-420 77316-422
3-Iodo-p-toluic acid 98%
3-Iodo-p-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₂
Melting Pt: 210…212 °C
Flash Pt: 168.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00010392
CAS Number: 82998-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR