3-Iodo-1-methyl-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
A263690-1G
A263690-25G
A263690-5G
A263690-100MG
A263690-10G
A263690-250MG
103812-636EA
17
USD
103812-636
103852-526
77092-172
77092-174
77092-176
77276-610
77276-612
77276-614
3-Iodo-1-methyl-1H-pyrazole 97%
3-Iodo-1-methyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₄H₅IN₂ Boiling Pt: 226.9 °C (760 mm Hg) Flash Pt: 91.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08435890 CAS Number: 92525-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...