3-Iodo-1H-pyrrolo[3,2-b]pyridine 98%
Supplier: Ambeed
|
Warning
|
A123705-250MG
A123705-1G
A123705-5G
103800-432EA
43
USD
103800-432
76810-668
77312-550
77312-552
3-Iodo-1H-pyrrolo[3,2-b]pyridine 98%
3-Iodo-1H-pyrrolo[3,2-b]pyridine
Precaución: for Research Use Only
Formula:
C₇H₅IN₂ MW: 244.032 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11559019 CAS Number: 1083181-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...