3-Iodo-1H-pyrazolo[3,4-b]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Iodo1H-pyrazolo[3,4-b]pyridine

A236030-100G A236030-10G A236030-1G A236030-5G
103811-062EA 14 USD
103811-062 103850-832 77031-198 77031-200 77089-864 77089-866
3-Iodo-1H-pyrazolo[3,4-b]pyridine 97%
3-Iodo-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄IN₃
MW: 245.02 g/mol
Boiling Pt: 379.8 °C (760 mm Hg)
Melting Pt: 177…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09749818
CAS Number: 117007-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR