3-Hydroxythietane 1,1-dioxide 95%

Supplier: Ambeed
Warning

A286456-1G A286456-5G A286456-100MG A286456-250MG
103813-714EA 86 USD
103813-714 103834-850 76837-386 76837-388 76988-582 77890-522
3-Hydroxythietane 1,1-dioxide 95%
3-Hydroxythietane 1,1-dioxide

Precaución: for Research Use Only

Formula: C₃H₆O₃S
Boiling Pt: 398.1 °C (760 mm Hg)
Flash Pt: 194.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22741759
CAS Number: 22524-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR