3-Hydroxypyridine-2-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxypicolinaldehyde

A138970-100MG A138970-10G A138970-250MG A138970-1G A138970-5G
103802-680EA 51 USD
103802-680 77285-638 77285-640 77285-642 77315-744 77315-746
3-Hydroxypyridine-2-carboxaldehyde 98%
3-Hydroxypyridine-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 292.3 °C (760 mm Hg)
Melting Pt: 80…82 °C
Flash Pt: 130.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06245505
CAS Number: 1849-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR