3-Hydroxypiperidine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Hydroxypiperidine Hydrochloride
A242744-1G
A242744-5G
A242744-100G
A242744-10G
103811-488EA
5
USD
103811-488
103851-308
77090-494
77090-496
77276-068
77276-070
3-Hydroxypiperidine hydrochloride 98%
3-Hydroxypiperidine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO MW: 137.61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012772 CAS Number: 64051-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...