3-Hydroxypyridine-2-carbonitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Cyano-3-hydroxypyridine, 3-Hydroxypicolinonitrile
A247913-1G
A247913-25G
A247913-100G
A247913-10G
A247913-250MG
A247913-5G
103811-800EA
7
USD
103811-800
103851-636
77090-972
77090-974
77276-242
77276-244
77276-246
77304-432
3-Hydroxypyridine-2-carbonitrile 98%
3-Hydroxypyridine-2-carbonitrile
Precaución: for Research Use Only
Formula:
C₆H₄N₂O MW: 120.1088 g/mol Boiling Pt: 429.2 °C (760 mm Hg) Melting Pt: 210…211 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01646135 CAS Number: 932-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...