3-Hydroxypicolinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxypyridine-2-carboxylic acid

A104849-100G A104849-100MG A104849-10G A104849-1G A104849-25G A104849-5G
103796-788EA 9 USD
103796-788 103822-514 77283-294 77283-296 77283-298 77283-300 77283-302 77309-374
3-Hydroxypicolinic acid 98%
3-Hydroxypicolinic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 213…218 °C
Flash Pt: 226.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00006294
CAS Number: 874-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR