3-Hydroxyisonicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxypyridine-4-carboxylic acid

A122337-100G A122337-25G A122337-1G
103800-206EA 6 USD
103800-206 77284-870 77284-872 77297-864
3-Hydroxyisonicotinic acid 97%
3-Hydroxyisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Boiling Pt: 519.3 °C (760 mm Hg)
Melting Pt: 317…319 °C
Flash Pt: 267.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00234165
CAS Number: 10128-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR