3-Hydroxyisobenzofuran-1(3H)-one 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxyphthalide, 3-Hydroxy-2-benzofuran-1(3H)-one, 3-Hydroxy-1(3H)-isobenzofuranone

A114644-100G A114644-25G A114644-5G A114644-250MG
103798-438EA 13 USD
103798-438 103824-282 77245-862 77245-864 77311-102 77745-848
3-Hydroxyisobenzofuran-1(3H)-one 97%
3-Hydroxyisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₈H₆O₃
Boiling Pt: 362.6 °C (760 mm Hg)
Flash Pt: 174 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00956142
CAS Number: 16859-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR