ɑ-Hydroxy-γ-butyrolactone 95%

Supplier: Ambeed
Warning

A237190-250MG A237190-10G
103811-138EA 17 USD
103811-138 103850-916 76692-204 77568-294
ɑ-Hydroxy-γ-butyrolactone 95%
alpha-Hydroxy-gamma-butyrolactone

Precaución: For research use only.

Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 249.3 °C (760 mm Hg)
Flash Pt: 122.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00134268
CAS Number: 19444-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR