3-Hydroxybenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: m-Hydroxybenzoic Acid

A233151-1000G A233151-100G A233151-500G A233151-25G
103809-642EA 34 USD
103809-642 77089-582 77113-062 77113-064 77303-954
3-Hydroxybenzoic acid 97%
3-Hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆O₃
Boiling Pt: 346.1 °C (760 mm Hg)
Melting Pt: 201 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002506
CAS Number: 99-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR