3-Hydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A256160-1000G A256160-25G A256160-100G A256160-10G A256160-500G
103809-818EA 128 USD
103809-818 77091-584 77091-586 77276-440 77276-442 77276-444
3-Hydroxybenzaldehyde 98%
3-Hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆O₂
MW: 122.12 g/mol
Boiling Pt: 240.9 °C (760 mm Hg)
Melting Pt: 100…103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003368
CAS Number: 100-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR