3'-Hydroxyacetophenone 98%

Supplier: Ambeed
Warning

A100185-1000G A100185-100G A100185-10G A100185-1G A100185-25G A100185-500G A100185-5G
103820-998EA 9 USD
103820-998 77148-142 77148-144 77148-146 77148-148 77148-150 77148-152 77296-588
3'-Hydroxyacetophenone 98%
3'-Hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 296 °C (760 mm Hg)
Melting Pt: 90…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002298
CAS Number: 121-71-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR