3-Hydroxy-5-(trifluoromethyl)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
A135215-100MG
A135215-250MG
A135215-1G
A135215-25G
A135215-10G
103802-112EA
12
USD
103802-112
77272-194
77272-196
77314-906
77314-908
77747-965
3-Hydroxy-5-(trifluoromethyl)benzoic acid 97%
3-Hydroxy-5-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₅F₃O₃ MW: 206.12 g/mol Boiling Pt: 333.7 °C (760 mm Hg) Melting Pt: 191…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368783 CAS Number: 328-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...