3-Hydroxy-4-nitrobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A357581-100G
A357581-10G
A357581-1G
A357581-25G
A357581-5G
103826-198EA
6
USD
103826-198
103837-202
76773-318
76773-320
76840-358
76840-360
76840-362
3-Hydroxy-4-nitrobenzaldehyde 98%
3-Hydroxy-4-nitrobenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅NO₄ Melting Pt: 127…131 °C Flash Pt: 138.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007109 CAS Number: 704-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...