3-Hydroxy-p-toluic acid 97%

Supplier: Ambeed
Warning

A145662-100G A145662-10G A145662-1G A145662-5G A145662-25G
103803-566EA 6 USD
103803-566 103841-648 77247-102 77247-104 77299-466 77316-956 77566-956
3-Hydroxy-p-toluic acid 97%
3-Hydroxy-p-toluic acid

Precaución: For research use only.

Formula: C₈H₈O₃
Boiling Pt: 352.6 °C (760 mm Hg)
Melting Pt: 205…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00002511
CAS Number: 586-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR