4,5-Dimethyl-3-hydroxy-2(5H)-furanone 95%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-4,5-dimethylfuran-2(5H)-one, 4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one

A107199-100G A107199-10G A107199-1G A107199-25G A107199-5G
103822-914EA 20 USD
103822-914 76994-998 76995-000 76995-002 76995-004 77283-556
4,5-Dimethyl-3-hydroxy-2(5H)-furanone 95%
4,5-Dimethyl-3-hydroxy-2(5H)-furanone

Precaución: for Research Use Only

Formula: C₆H₈O₃
MW: 128.13 g/mol
Boiling Pt: 312 °C (760 mm Hg)
Melting Pt: 26…29 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059957
CAS Number: 28664-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR