DL-Pantolactone 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, DL-Pantoyl Lactone, 3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one, DL-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone
A229075-100G
A229075-10G
A229075-500G
A229075-1000G
A229075-25G
A229075-5G
103809-622EA
247
USD
103809-622
103850-346
76801-044
76801-046
76801-048
77303-844
77303-846
77303-848
DL-Pantolactone 96%
DL-Pantolactone
Precaución: for Research Use Only
Formula:
C₆H₁₀O₃ MW: 130.14 g/mol Boiling Pt: 224.6 °C (760 mm Hg) Melting Pt: 74…78 °C Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064333 CAS Number: 79-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...