3-Hydroxy-3-phenylpropanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-3-phenylpropionic acid

A272969-1G A272969-25G A272969-100G A272969-250MG
103813-118EA 11 USD
103813-118 103853-034 77092-824 77092-826 77276-814 77748-523
3-Hydroxy-3-phenylpropanoic acid 98%
3-Hydroxy-3-phenylpropanoic acid

Precaución: for Research Use Only

Formula: C9H10O3
MW: 166.1700 g/mol
Boiling Pt: 329.4 °C (760 mm Hg)
Flash Pt: 167.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00137907
CAS Number: 3480-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR