3-Hydroxy-3-methylhexanoic acid 96%

Supplier: Ambeed
Warning

A149671-100MG A149671-250MG A149671-1G A149671-5G
103804-068EA 132 USD
103804-068 103842-194 77272-936 77272-938 77317-696 77317-698
3-Hydroxy-3-methylhexanoic acid 96%
3-Hydroxy-3-methylhexanoic acid

Precaución: for Research Use Only

Formula: C₇H₁₄O₃
MW: 146.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779366
CAS Number: 58888-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR