DL-Tropic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-2-phenylpropanoic acid, Tropic acid, 3-Hydroxy-2-phenylpropionic acid, (±)-Tropic acid, 2-Phenylhydracrylic acid

A179976-1000G A179976-25G A179976-100G A179976-10G A179976-5G A179976-500G
103809-138EA 347 USD
103809-138 77083-322 77083-324 77111-386 77111-388 77111-390 77685-408
DL-Tropic acid 98%
DL-Tropic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 322.5 °C (760 mm Hg)
Melting Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00004255
CAS Number: 529-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR