3-Hydroxy-2-nitrobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A450286-1G
A450286-10G
A450286-25G
103827-786EA
16
USD
103827-786
103838-856
76842-672
77143-518
77307-358
3-Hydroxy-2-nitrobenzoic acid 97%
3-Hydroxy-2-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₅NO₅ MW: 183.12 g/mol Boiling Pt: 362.9 °C (760 mm Hg) Melting Pt: 179…181 °C Flash Pt: 167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017003 CAS Number: 602-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...