3-Hydroxy-2-naphthoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Hydroxy-naphthalene-2-carboxylic acid, 2-Hydroxy-3-naphthoic acid
A274878-100G
A274878-25G
A274878-500G
103809-904EA
31
USD
103809-904
76875-168
77092-998
77593-696
3-Hydroxy-2-naphthoic acid 98%
3-Hydroxy-2-naphthoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₈O₃ MW: 188.18 g/mol Melting Pt: 218…221 °C Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004103 CAS Number: 92-70-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...