3-Hydroxy-2-methylbenzonitrile 95%

Supplier: Ambeed
Warning

A235823-1G A235823-100MG
103811-036EA 162 USD
103811-036 103850-804 77089-822 77275-892
3-Hydroxy-2-methylbenzonitrile 95%
3-Hydroxy-2-methylbenzonitrile

Precaución: for Research Use Only

Formula: C8H7NO
MW: 133.1473 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683035
CAS Number: 55289-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR