3-Hydroxy-2H-pyran-2-one 97%

Supplier: Ambeed
Warning

A120108-100MG A120108-1G A120108-250MG
103799-882EA 138 USD
103799-882 77271-442 77297-734 77571-202
3-Hydroxy-2H-pyran-2-one 97%
3-Hydroxy-2H-pyran-2-one

Precaución: for Research Use Only

Formula: C₅H₄O₃
Storage Temperature: Ambient
MDL Number: MFCD01318559
CAS Number: 496-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR