3-Hydrazinylpyridine dihydrochloride 97%

Supplier: Ambeed
Warning

A293417-1G A293417-100MG A293417-10G A293417-250MG A293417-25G A293417-5G
103813-980EA 19 USD
103813-980 103835-118 76837-736 77277-208 77277-210 77277-212 77305-626 77305-628
3-Hydrazinylpyridine dihydrochloride 97%
3-Hydrazinylpyridine dihydrochloride

Precaución: for Research Use Only

Formula: C5H9Cl2N3
MW: 182.0511 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08576202
CAS Number: 364727-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR