3H-Imidazo[4,5-c]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Aza-1H-benzoimidazole, 5-Azabenzimidazole

A101883-10G A101883-250MG A101883-1G A101883-5G A101883-25G
103796-194EA 13 USD
103796-194 103821-888 77270-268 77270-270 77308-774 77308-776 77747-759
3H-Imidazo[4,5-c]pyridine 98%
3H-Imidazo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₅N₃
MW: 119.13 g/mol
Boiling Pt: 425.1 °C (760 mm Hg)
Melting Pt: 168…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00051808
CAS Number: 272-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR