3H-Imidazo[4,5-b]pyridine-6-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 6-Carboxy-3H-imidazo[4,5-b]pyridine

A217795-100MG A217795-250MG
103818-658EA 200 USD
103818-658 103849-386 76975-742 76975-744
3H-Imidazo[4,5-b]pyridine-6-carboxylic acid 97%
3H-Imidazo[4,5-b]pyridine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06254751
CAS Number: 24638-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR